Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194986.1 | NP_001181915.1 | 7 | 3865 | ||
XM_006710640.3 | XP_006710703.1 | 8 | 4446 | |||
XM_017001260.1 | XP_016856749.1 | 7 | 4176 | |||
XM_017001261.1 | XP_016856750.1 | 5 | 3896 | |||
XM_017001263.1 | XP_016856752.1 | 5 | 4315 | |||
GenBank mRNA | AK090467.1 | - | 1 | 1305 |