Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321829.1 | NP_001308758.1 | 17 | 6729 | ||
NM_018017.3 | NP_060487.2 | 16 | 6604 | |||
NR_135815.1 | 16 | 7216 | ||||
XM_011539915.2 | XP_011538217.1 | 15 | 5947 | |||
GenBank mRNA | AK024739.1 | - | 1 | 1052 | ||
AK094498.1 | - | 1 | 2092 |