Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107983998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527018.2 | XP_011525320.1 | 1 | 98 | ||
XM_011527021.1 | XP_011525323.1 | 1 | 98 |
Gene Symbol: ZNF581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026867.1 | XP_016882356.1 | 1 | 65 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK055093.1 | - | 1 | 90 | Chr. 19 - 55635003 - 55637234 [+] on Build GRCh38 |