Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001973.3 | NP_001964.2 | 5 | 8367 | ||
GenBank mRNA | AK024944.1 | - | 1 | 1288 |