Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322080.1 | NP_001309009.1 | 6 | 4774 | ||
NM_001322081.1 | NP_001309010.1 | 6 | 4675 | |||
NM_032718.4 | NP_116107.3 | 6 | 4658 | |||
NR_136173.1 | 7 | 4742 | ||||
NR_136174.1 | 7 | 2479 | ||||
XM_011512005.2 | XP_011510307.1 | 6 | 4781 | |||
XM_011512006.2 | XP_011510308.1 | 6 | 4598 | |||
XM_011512007.2 | XP_011510309.1 | 4 | 4255 | |||
GenBank mRNA | U79290.1 | - | 1 | 1180 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK821567.1 | - | 1 | 113 | Chr. 2 - 102714809 - 102715377 [-] on Build GRCh38 |