Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521985.1 | XP_011520287.1 | 27 | 7054 | ||
XM_017022533.1 | XP_016878022.1 | 29 | 7309 | |||
GenBank mRNA | AK125173.1 | - | 1 | 68 |