Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266650.3 | XP_005266707.1 | 4 | 8794 | ||
XM_005266652.3 | XP_005266709.1 | 2 | 8587 | |||
XM_011525878.2 | XP_011524180.1 | 4 | 8925 | |||
XM_011525879.2 | XP_011524181.1 | 5 | 9105 | |||
XM_017025612.1 | XP_016881101.1 | 3 | 8710 | |||
XM_017025613.1 | XP_016881102.1 | 4 | 8911 | |||
GenBank mRNA | BC071787.1 | - | 1 | 1438 |