Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC284570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040156.1 | - | 1 | 94 |
Gene Symbol: GPR37L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510158.2 | XP_011508460.1 | 2 | 2594 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG910726.1 | - | 1 | 94 | Chr. 1 - 202130056 - 202130905 [+] on Build GRCh38 |