Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153686.8 | NP_710153.2 | 7 | 4093 | ||
NR_136669.1 | 5 | 3919 | ||||
XM_011513821.2 | XP_011512123.1 | 9 | 8824 | |||
XM_011513822.2 | XP_011512124.1 | 8 | 8797 | |||
XM_011513823.2 | XP_011512125.1 | 8 | 8670 | |||
XM_011513826.2 | XP_011512128.1 | 6 | 3877 | |||
XM_017007961.1 | XP_016863450.1 | 8 | 10106 | |||
XM_017007962.1 | XP_016863451.1 | 8 | 8748 | |||
XM_017007963.1 | XP_016863452.1 | 8 | 9051 | |||
GenBank mRNA | AL133031.1 | - | 1 | 2222 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA062154.1 | - | 1 | 227 | Chr. 4 - 17842380 - 17842934 [-] on Build GRCh38 |