Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153686.8 | NP_710153.2 | 7 | 3632 | ||
NR_136669.1 | 5 | 3458 | ||||
XM_011513821.2 | XP_011512123.1 | 9 | 8363 | |||
XM_011513822.2 | XP_011512124.1 | 8 | 8336 | |||
XM_011513823.2 | XP_011512125.1 | 8 | 8209 | |||
XM_011513826.2 | XP_011512128.1 | 6 | 3416 | |||
XM_017007961.1 | XP_016863450.1 | 8 | 9645 | |||
XM_017007962.1 | XP_016863451.1 | 8 | 8287 | |||
XM_017007963.1 | XP_016863452.1 | 8 | 8590 | |||
GenBank mRNA | AL133031.1 | - | 1 | 1761 |