Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271208.2 | XP_005271265.1 | 20 | 6990 | ||
XM_017002347.1 | XP_016857836.1 | 22 | 7199 | |||
XM_017002349.1 | XP_016857838.1 | 20 | 6954 | |||
GenBank mRNA | BC042023.1 | - | 1 | 2313 |