Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324266.1 | NP_001311195.1 | 7 | 4222 | ||
NM_024576.4 | NP_078852.3 | 7 | 4219 | |||
XM_011536117.2 | XP_011534419.1 | 7 | 4155 | |||
XM_011536118.2 | XP_011534420.1 | 7 | 4004 | |||
XM_017011303.1 | XP_016866792.1 | 7 | 3784 | |||
XM_017011304.1 | XP_016866793.1 | 7 | 3781 | |||
XM_017011305.1 | XP_016866794.1 | 7 | 3677 | |||
XM_017011306.1 | XP_016866795.1 | 7 | 4707 | |||
GenBank mRNA | AK124833.1 | - | 1 | 1959 |