Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278199.1 | NP_001265128.1 | 15 | 6505 | ||
NM_003100.3 | NP_003091.2 | 15 | 5954 | |||
GenBank mRNA | AK026718.1 | - | 1 | 1579 |