Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272600.3 | XP_005272657.1 | 5 | 10396 | ||
XM_011543899.2 | XP_011542201.1 | 5 | 12002 | |||
XM_011543900.2 | XP_011542202.1 | 6 | 10457 | |||
XM_017029486.1 | XP_016884975.1 | 6 | 10515 | |||
XM_017029487.1 | XP_016884976.1 | 5 | 10748 | |||
GenBank mRNA | AK026716.1 | - | 1 | 1017 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA571914.1 | - | 1 | 795 | Chr. X - 47923941 - 47924804 [+] on Build GRCh38 |