Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004531.4 | NP_004522.1 | 7 | 2428 | ||
NM_176806.3 | NP_789776.1 | 7 | 1967 | |||
GenBank mRNA | AK095832.1 | - | 1 | 677 |