Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002072.4 | NP_002063.2 | 7 | 5710 | ||
GenBank mRNA | AK055619.1 | - | 1 | 1537 |