Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARGC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513770.2 | XP_011512072.1 | 1 | 195 | ||
GenBank mRNA | HQ695733.1 | - | 1 | 195 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK095100.1 | - | 1 | 876 | Chr. 4 - 23879202 - 23881973 [-] on Build GRCh38 |