Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281293.1 | NP_001268222.1 | 2 | 4682 | ||
NM_001281294.1 | NP_001268223.1 | 3 | 4574 | |||
NM_012482.4 | NP_036614.1 | 2 | 4678 | |||
XM_017000887.1 | XP_016856376.1 | 5 | 6063 | |||
XM_017000888.1 | XP_016856377.1 | 5 | 5021 | |||
GenBank mRNA | BC016000.1 | - | 1 | 950 |