Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 5888 | ||
GenBank mRNA | AK024927.1 | - | 1 | 1373 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM786734.1 | - | 1 | 410 | Chr. 6 - 54944335 - 54944998 [+] on Build GRCh38 |