Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123567.1 | - | 1 | 1387 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM285393.1 | - | 1 | 395 | Chr. 11 - 35197162 - 35198555 [+] on Build GRCh38 |