Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001561.5 | NP_001552.2 | 9 | 2847 | ||
XM_011541386.2 | XP_011539688.1 | 8 | 2862 | |||
GenBank mRNA | AK125490.1 | - | 1 | 865 |