Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517379.2 | XP_011515681.1 | 21 | 3196 | ||
XM_017013983.1 | XP_016869472.1 | 20 | 3001 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK057260.1 | - | 1 | 1218 | Chr. 2 - 74196704 - 74198560 [-] on Build GRCh38 |