Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC102B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526174.2 | XP_011524476.1 | 9 | 7207 | ||
GenBank mRNA | AK026731.1 | - | 1 | 150 |