Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521005.2 | XP_011519307.1 | 1 | 613 | ||
GenBank mRNA | AY231477.1 | - | 1 | 191 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA632847.1 | - | 1 | 210 | Chr. 12 - 851424 - 859375 [+] on Build GRCh38 |