Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000043.5 | NP_000034.1 | 9 | 3340 | ||
NM_001320619.1 | NP_001307548.1 | 8 | 3257 | |||
NM_152871.3 | NP_690610.1 | 8 | 3277 | |||
NM_152872.3 | NP_690611.1 | 8 | 3315 | |||
NR_028033.3 | 7 | 3168 | ||||
NR_028034.3 | 6 | 3030 | ||||
NR_028035.3 | 7 | 3093 | ||||
NR_028036.3 | 8 | 3231 | ||||
NR_135313.1 | 7 | 3148 | ||||
NR_135314.1 | 9 | 3331 | ||||
NR_135315.1 | 7 | 3084 | ||||
GenBank mRNA | AK026195.1 | - | 1 | 629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX093666.1 | - | 1 | 395 | Chr. 10 - 89016070 - 89016855 [+] on Build GRCh38 |