Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096543.1 | - | 1 | 2565 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK299173.1 | - | 1 | 259 | Chr. 6 - 3230246 - 3230747 [+] on Build GRCh38 |