Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001182.4 | NP_001173.2 | 18 | 4778 | ||
NM_001201377.1 | NP_001188306.1 | 18 | 4778 | |||
NM_001202404.1 | NP_001189333.2 | 16 | 4586 | |||
GenBank mRNA | AK123774.1 | - | 1 | 1136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX099612.1 | - | 3 | 592 | Chr. 5 - 126541956 - 126747460 [-] on Build GRCh38 |