Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 10 | 4364 | ||
NM_001257159.1 | NP_001244088.1 | 9 | 4316 | |||
NM_018718.2 | NP_061188.1 | 11 | 4580 | |||
NR_046443.1 | 11 | 4504 | ||||
GenBank mRNA | BC047500.1 | - | 1 | 2081 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK001131.1 | - | 1 | 514 | Chr. 7 - 130395535 - 130396189 [-] on Build GRCh38 |