Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127909.1 | - | 2 | 328 |
Gene Symbol: UBL7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW081069.1 | - | 1 | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA940586.1 | - | 2 | 328 | Chr. 15 - 74480863 - 74490286 [-] on Build GRCh38 |