Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127902.1 | - | 1 | 1394 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK005511.1 | - | 1 | 24 | Chr. X - 41335980 - 41336636 [+] on Build GRCh38 |