Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184763.1 | NP_001171692.1 | 8 | 6029 | ||
NM_144624.2 | NP_653225.2 | 7 | 6281 | |||
NM_175866.4 | NP_787062.1 | 8 | 6370 | |||
GenBank mRNA | AK026668.1 | - | 1 | 217 | ||
AL137257.1 | - | 1 | 2379 | |||
BC001977.1 | - | 1 | 1102 | |||
BC043436.2 | - | 1 | 2095 |