Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF085965.1 | - | 1 | 295 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | H72029.1 | - | 1 | 295 | Chr. 17 - 48855803 - 48856231 [+] on Build GRCh38 |