Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542397.2 | XP_011540699.1 | 1 | 186 | ||
XM_011542398.2 | XP_011540700.1 | 1 | 191 | |||
GenBank mRNA | AK098790.1 | - | 1 | 219 |