Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013537.1 | XP_016869026.1 | 7 | 1973 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK130667.1 | - | 1 | 2104 | Chr. 8 - 27717072 - 27719537 [+] on Build GRCh38 |