Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 5492 | ||
NM_020640.3 | NP_065691.2 | 7 | 5474 | |||
XM_011512912.1 | XP_011511214.1 | 7 | 809 | |||
XM_011512914.1 | XP_011511216.1 | 7 | 848 | |||
XM_011512915.2 | XP_011511217.1 | 8 | 1370 | |||
XM_011512916.2 | XP_011511218.1 | 7 | 1298 | |||
XM_017006633.1 | XP_016862122.1 | 6 | 725 | |||
XM_017006634.1 | XP_016862123.1 | 7 | 814 | |||
GenBank mRNA | AK095773.1 | - | 1 | 178 |