Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 7561 | ||
NM_020640.3 | NP_065691.2 | 7 | 7543 | |||
XM_011512912.1 | XP_011511214.1 | 7 | 2878 | |||
XM_011512914.1 | XP_011511216.1 | 7 | 2917 | |||
XM_011512915.2 | XP_011511217.1 | 8 | 3439 | |||
XM_011512916.2 | XP_011511218.1 | 7 | 3367 | |||
XM_017006633.1 | XP_016862122.1 | 7 | 2689 | |||
XM_017006634.1 | XP_016862123.1 | 7 | 2883 | |||
GenBank mRNA | AK095773.1 | - | 1 | 2247 |