Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124599.1 | - | 1 | 2812 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV725837.1 | - | 1 | 227 | Chr. 13 - 100604856 - 100605553 [-] on Build GRCh38 |