Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100130744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046285.1 | 1 | 2026 | |||
GenBank mRNA | AF289590.1 | - | 1 | 2046 | ||
AK129556.1 | - | 1 | 40 |
Gene Symbol: OTULIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514151.2 | XP_011512453.1 | 8 | 7135 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX507119.1 | - | 1 | 88 | Chr. 5 - 14714632 - 14715242 [+] on Build GRCh38 |