Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309192.1 | NP_001296121.1 | 14 | 6781 | ||
NM_001309193.1 | NP_001296122.1 | 17 | 6862 | |||
NM_004442.7 | NP_004433.2 | 16 | 6958 | |||
NM_017449.4 | NP_059145.2 | 16 | 6955 | |||
XM_006710441.3 | XP_006710504.1 | 16 | 7051 | |||
XM_006710442.3 | XP_006710505.1 | 16 | 6933 | |||
GenBank mRNA | AK125423.1 | - | 1 | 738 |
Gene Symbol: TMEM60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095664.1 | - | 2 | 345 |