Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062445.1 | - | 1 | 501 | ||
BC150293.1 | - | 20 | 3826 | 8860318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU172356.1 | - | 1 | 144 | Chr. 1 - 203733342 - 203734153 [+] on Build GRCh38 |