Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038970.1 | 1 | 2526 | |||
GenBank mRNA | AK092333.1 | - | 1 | 1378 | ||
BC035145.1 | - | 1 | 2529 |