Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038970.1 | 1 | 3078 | |||
GenBank mRNA | AK092333.1 | - | 1 | 1930 | ||
AK093962.1 | - | 1 | 268 | |||
BC035145.1 | - | 1 | 3081 |