Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF278605.1 | - | 1 | 124 | ||
AK128047.1 | - | 1 | 2849 | |||
BC045722.1 | - | 4 | 1068 |