Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006112.3 | NP_006103.1 | 10 | 4025 | ||
NR_036543.1 | 10 | 4017 | ||||
NR_036544.1 | 10 | 3978 |