Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006112.3 | NP_006103.1 | 10 | 4280 | ||
NR_036543.1 | 10 | 4272 | ||||
NR_036544.1 | 10 | 4233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ187153.1 | - | 1 | 503 | Chr. 1 - 39756172 - 39756771 [+] on Build GRCh38 |