Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120424.1 | 3 | 371 | |||
GenBank mRNA | AK093174.1 | - | 3 | 371 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB025349.1 | - | 3 | 371 | Chr. 13 - 45106279 - 45112002 [+] on Build GRCh38 |