Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142386.2 | NP_001135858.1 | 12 | 12231 | ||
GenBank mRNA | BC038512.2 | - | 1 | 4033 |