Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC14A2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110899.1 | 5 | 485 | |||
GenBank mRNA | AK126075.1 | - | 5 | 485 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB091714.1 | - | 5 | 485 | Chr. 18 - 45483338 - 45507036 [-] on Build GRCh38 |