Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321305.1 | NP_001308234.1 | 6 | 2152 | ||
XM_011532541.2 | XP_011530843.1 | 8 | 2504 | |||
XM_011532543.2 | XP_011530845.1 | 9 | 2788 | |||
XM_011532545.2 | XP_011530847.1 | 8 | 1212 | |||
XM_017003377.1 | XP_016858866.1 | 7 | 1110 | |||
XM_017003378.1 | XP_016858867.1 | 9 | 2463 | |||
GenBank mRNA | AK091998.1 | - | 1 | 1680 |