Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522462.2 | XP_011520764.1 | 4 | 4278 | ||
GenBank mRNA | AK057657.1 | - | 1 | 1642 | ||
AK095835.1 | - | 1 | 3007 | |||
AK123562.1 | - | 1 | 2506 | |||
AL832650.1 | - | 1 | 4111 |